Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214040.1 | NP_999205.1 | 13 | 1349 | 99 | |
XM_021093506.1 | XP_020949165.1 | 1400 | 99 | |||
XM_021093507.1 | XP_020949166.1 | 1352 | 99 | |||
XM_021093508.1 | XP_020949167.1 | 1434 | 99 | |||
XM_021093509.1 | XP_020949168.1 | 1466 | 99 | |||
XM_021093510.1 | XP_020949169.1 | 1418 | 99 | |||
XM_021093511.1 | XP_020949170.1 | 1400 | 99 | |||
XM_021093512.1 | XP_020949171.1 | 1352 | 99 | |||
GenBank mRNA | AK232889.1 | - | 13 | 1353 | 99 | |
AK238766.1 | - | 13 | 1306 | 99 | ||
U00794.1 | - | 13 | 1349 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |