Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214182.1 | NP_999347.1 | 5 - 6 | 490 | 78 | |
GenBank mRNA | L02900.1 | - | 5 - 6 | 490 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |