Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214341.1 | NP_999506.1 | 9 - 10 | 1098 | 97 | |
GenBank mRNA | AK395683.1 | - | 9 - 10 | 1143 | 97 | |
AY266142.1 | - | 9 - 10 | 1098 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |