Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130730.1 | NP_001124202.1 | 7 - 8 | 710 | 92 | |
GenBank mRNA | AB529537.1 | - | 6 - 7 | 687 | 92 | |
AK238335.1 | - | 6 - 7 | 909 | 92 | ||
AK392822.1 | - | 7 - 8 | 704 | 92 | ||
EF530130.1 | - | 7 - 8 | 690 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |