Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159659.1 | NP_001153131.1 | 1 - 2 | 50 | 101 | |
XM_013997109.2 | XP_013852563.1 | 116 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |