Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184895.1 | NP_001171824.1 | 7 - 8 | 1001 | 64 | |
GenBank mRNA | AK235245.1 | - | 7 - 8 | 703 | 64 | |
AK350347.1 | - | 7 - 8 | 702 | 64 | ||
AK392413.1 | - | 7 - 8 | 706 | 64 | ||
AK393586.1 | - | 7 - 8 | 708 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |