Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185165.1 | NP_001172094.1 | 2 - 3 | 212 | 104 | |
XM_005654358.3 | XP_005654415.1 | 251 | 104 | |||
GenBank mRNA | AK344714.1 | - | 2 - 3 | 208 | 104 | |
AY609842.1 | - | 2 - 3 | 197 | 104 | ||
BF075005.1 | - | 2 - 3 | 199 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |