Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185171.1 | NP_001172100.1 | 3 - 4 | 329 | 70 | |
XM_005671491.3 | XP_005671548.1 | 165 | 70 | |||
XM_005671494.3 | XP_005671551.1 | 110 | 70 | |||
GenBank mRNA | AK234444.1 | - | 3 - 4 | 328 | 70 | |
AY609511.1 | - | 2 - 3 | 140 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |