Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185836.1 | NP_001172765.1 | 3 - 4 | 153 | 80 | |
XM_005656377.3 | XP_005656434.1 | 483 | 80 | |||
XM_005656378.3 | XP_005656435.1 | 481 | 80 | |||
GenBank mRNA | AK231110.1 | - | 3 - 4 | 153 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |