Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190176.1 | NP_001177105.1 | 5 - 6 | 899 | 93 | |
XM_005654380.3 | XP_005654437.1 | 3526 | 93 | |||
XM_013984860.2 | XP_013840314.1 | 3526 | 93 | |||
GenBank mRNA | AK232190.1 | - | 5 - 6 | 899 | 93 | |
AK345725.1 | - | 5 - 6 | 899 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |