Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190181.1 | NP_001177110.1 | 1 - 2 | 213 | 92 | |
XM_021098029.1 | XP_020953688.1 | 206 | 92 | |||
GenBank mRNA | AK231349.1 | - | 1 - 2 | 131 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |