Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195359.1 | NP_001182288.1 | 3 - 4 | 475 | 99 | |
XM_005668865.3 | XP_005668922.1 | 959 | 99 | |||
XM_005668866.3 | XP_005668923.1 | 960 | 99 | |||
XM_005668867.3 | XP_005668924.1 | 485 | 99 | |||
GenBank mRNA | AK237413.1 | - | 3 - 4 | 489 | 99 | |
AK348542.1 | - | 3 - 4 | 427 | 99 | ||
GU373686.1 | - | 3 - 4 | 475 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |