Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190287.1 | NP_001177216.1 | 4 | 749 | 67 | |
GenBank mRNA | AK232622.1 | - | 4 | 883 | 67 | |
AK345988.1 | - | 4 | 883 | 67 | ||
HM046513.1 | - | 4 | 749 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |