Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172368.1 | NP_001165839.1 | 2 - 3 | 213 | 66 | |
GenBank mRNA | AY610264.1 | - | 1 - 2 | 147 | 66 | |
FJ494917.1 | - | 2 - 3 | 213 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |