Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190280.1 | NP_001177209.1 | 8 - 9 | 2063 | 72 | |
XM_021078467.1 | XP_020934126.1 | 2452 | 72 | |||
GenBank mRNA | HM046519.1 | - | 8 - 9 | 2063 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |