Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195356.1 | NP_001182285.1 | 2 - 3 | 350 | 107 | |
GenBank mRNA | AK240357.1 | - | 3 - 4 | 411 | 107 | |
GU373666.1 | - | 2 - 3 | 350 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |