Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243457.1 | NP_001230386.1 | 1 - 2 | 147 | 84 | |
GenBank mRNA | AK352479.1 | - | 1 - 2 | 147 | 84 | |
BE231521.1 | - | 1 - 2 | 141 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |