Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243486.1 | NP_001230415.1 | 2 - 3 | 205 | 77 | |
XM_005663352.3 | XP_005663409.1 | 249 | 77 | |||
XM_013997041.2 | XP_013852495.1 | 255 | 77 | |||
GenBank mRNA | AK348954.1 | - | 2 - 3 | 202 | 77 | |
AY610287.1 | - | 2 - 3 | 210 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |