Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243498.1 | NP_001230427.1 | 2 - 3 | 353 | 103 | |
XM_005670259.3 | XP_005670316.2 | 407 | 103 | |||
GenBank mRNA | AK343803.1 | - | 2 - 3 | 353 | 103 | |
AK392457.1 | - | 2 - 3 | 348 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |