Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243664.1 | NP_001230593.1 | 5 - 6 | 649 | 103 | |
XM_013992739.2 | XP_013848193.1 | 466 | 103 | |||
XM_021101116.1 | XP_020956775.1 | 599 | 103 | |||
XM_021101239.1 | XP_020956898.1 | 410 | 103 | |||
GenBank mRNA | AK345353.1 | - | 5 - 6 | 649 | 103 | |
DY419200.1 | - | 5 - 6 | 582 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |