Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243896.1 | NP_001230825.1 | 7 - 8 | 803 | 100 | |
GenBank mRNA | AK234631.1 | - | 7 - 8 | 797 | 100 | |
AK238735.1 | - | 7 - 8 | 803 | 100 | ||
AK347564.1 | - | 7 - 8 | 797 | 100 | ||
AK392606.1 | - | 7 - 8 | 794 | 100 | ||
AY609618.1 | - | 7 - 8 | 777 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |