Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244077.1 | NP_001231006.1 | 4 - 5 | 495 | 63 | |
GenBank mRNA | AK236531.1 | - | 3 - 4 | 358 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |