Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244149.1 | NP_001231078.1 | 11 - 12 | 1300 | 76 | |
XM_021066089.1 | XP_020921748.1 | 1174 | 76 | |||
GenBank mRNA | AK234845.1 | - | 10 - 11 | 1071 | 76 | |
AK239091.1 | - | 11 - 12 | 1296 | 76 | ||
AK395140.1 | - | 10 - 11 | 1157 | 76 | ||
AK396275.1 | - | 11 - 12 | 1572 | 76 | ||
CK459694.1 | - | 8 - 9 | 668 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |