Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244241.1 | NP_001231170.1 | 7 - 8 | 794 | 104 | |
XM_005659114.3 | XP_005659171.1 | 813 | 104 | |||
GenBank mRNA | AK233198.1 | - | 7 - 8 | 768 | 104 | |
AK237427.1 | - | 8 - 9 | 675 | 104 | ||
AK351349.1 | - | 7 - 8 | 794 | 104 | ||
AK391167.1 | - | 7 - 8 | 778 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |