Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244396.1 | NP_001231325.1 | 5 - 6 | 548 | 93 | |
XM_005667397.2 | XP_005667454.2 | 593 | 93 | |||
GenBank mRNA | AK350554.1 | - | 5 - 6 | 548 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |