Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244398.1 | NP_001231327.1 | 6 - 7 | 652 | 122 | |
GenBank mRNA | AK236324.1 | - | 6 - 7 | 652 | 122 | |
AK393624.1 | - | 6 - 7 | 685 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |