Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244491.1 | NP_001231420.1 | 2 - 3 | 724 | 103 | |
GenBank mRNA | AK231741.1 | - | 2 - 3 | 729 | 103 | |
AK236121.1 | - | 2 - 3 | 724 | 103 | ||
AK350390.1 | - | 2 - 3 | 728 | 103 | ||
AY609562.1 | - | 2 - 3 | 330 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |