Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244493.1 | NP_001231422.1 | 3 - 4 | 331 | 98 | |
XM_021074514.1 | XP_020930173.1 | 657 | 98 | |||
GenBank mRNA | AK343867.1 | - | 3 - 4 | 382 | 98 | |
AY609907.1 | - | 3 - 4 | 335 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |