Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVRIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244699.1 | NP_001231628.1 | 2 - 3 | 1161 | 104 | |
XM_005661897.3 | XP_005661954.1 | 808 | 104 | |||
XM_021085659.1 | XP_020941318.1 | 826 | 104 | |||
GenBank mRNA | AK240369.1 | - | 2 - 3 | 1161 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |