Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244776.1 | NP_001231705.1 | 2 - 3 | 297 | 88 | |
GenBank mRNA | AK351235.1 | - | 2 - 3 | 230 | 88 | |
AK395110.1 | - | 2 - 3 | 296 | 88 | ||
AK400544.1 | - | 2 - 3 | 230 | 88 | ||
BP145298.1 | - | 2 - 3 | 297 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |