Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244831.1 | NP_001231760.1 | 3 - 4 | 669 | 81 | |
GenBank mRNA | AK235372.1 | - | 3 - 4 | 669 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |