Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VVSUC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF021808.1 | - | 1 - 2 | 523 | 65 | |
AF182445.1 | - | 1 - 2 | 493 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Vitis vinifera Probe spans exons |