Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cstf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001220.1 | NP_001001220.1 | 3 - 4 | 610 | 87 | |
GenBank mRNA | BC067934.1 | - | 3 - 4 | 610 | 87 | |
CT030488.1 | - | 2 - 3 | 474 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |