Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002904.2 | NP_001002904.1 | 2 - 3 | 1677 | 67 | |
GenBank mRNA | AJ784816.1 | - | 2 - 3 | 1721 | 67 | |
BC167262.1 | - | 2 - 3 | 1530 | 67 | ||
BC171316.1 | - | 2 - 3 | 1641 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |