Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003921.1 | NP_001003921.1 | 4 - 5 | 536 | 87 | |
GenBank mRNA | BC062521.1 | - | 4 - 5 | 526 | 87 | |
BC079960.1 | - | 4 - 5 | 536 | 87 | ||
CR761010.2 | - | 601 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |