Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dscc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004834.1 | NP_001004834.1 | 2 - 3 | 255 | 77 | |
GenBank mRNA | BC074630.1 | - | 2 - 3 | 255 | 77 | |
CR760288.2 | - | 2 - 3 | 262 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |