Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004888.1 | NP_001004888.1 | 3 - 4 | 368 | 103 | |
XM_018096198.1 | XP_017951687.1 | 3 - 4 | 405 | 103 | ||
GenBank mRNA | BC075290.1 | - | 3 - 4 | 368 | 103 | |
CR761259.2 | - | 3 - 4 | 397 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |