Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004981.1 | NP_001004981.1 | 4 - 5 | 512 | 124 | |
XM_018092030.1 | XP_017947519.1 | 3 - 4 | 477 | 124 | ||
GenBank mRNA | BC075530.1 | - | 4 - 5 | 512 | 124 | |
CR762186.2 | - | 4 - 5 | 460 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |