Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trmt61a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005009.1 | NP_001005009.1 | 3 - 4 | 712 | 80 | |
GenBank mRNA | BC076660.1 | - | 3 - 4 | 712 | 80 | |
CR762175.2 | - | 3 - 4 | 722 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |