Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203534.1 | NP_988865.1 | 4 - 5 | 608 | 89 | |
GenBank mRNA | AY333424.1 | - | 4 - 5 | 608 | 89 | |
BC170803.1 | - | 4 - 5 | 599 | 89 | ||
BC170805.1 | - | 4 - 5 | 599 | 89 | ||
CR926370.2 | - | 4 - 5 | 584 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |