Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203619.1 | NP_988950.1 | 1 - 2 | 428 | 61 | |
GenBank mRNA | BC061325.1 | - | 1 - 2 | 428 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |