Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203665.1 | NP_988996.1 | 5 - 6 | 722 | 88 | |
GenBank mRNA | BC061269.1 | - | 5 - 6 | 722 | 88 | |
BC122046.1 | - | 5 - 6 | 710 | 88 | ||
CU075322.1 | - | 5 - 6 | 690 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |