Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203701.1 | NP_989032.1 | 1 - 2 | 473 | 98 | |
GenBank mRNA | BC061394.1 | - | 1 - 2 | 473 | 98 | |
CR761582.2 | - | 1 - 2 | 174 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |