Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203716.1 | NP_989047.1 | 3 - 4 | 500 | 104 | |
XM_012962836.2 | XP_012818290.1 | 3 - 4 | 635 | 104 | ||
GenBank mRNA | BC061354.1 | - | 3 - 4 | 500 | 104 | |
CR761467.2 | - | 3 - 4 | 367 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |