Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203775.1 | NP_989106.1 | 1 - 2 | 119 | 57 | |
XM_012970259.2 | XP_012825713.1 | 1 - 2 | 197 | 57 | ||
GenBank mRNA | BC062497.1 | - | 1 - 2 | 119 | 57 | |
CR762193.2 | - | 1 - 2 | 91 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |