Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psma6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203782.1 | NP_989113.1 | 1 - 2 | 90 | 104 | |
GenBank mRNA | BC061438.1 | - | 1 - 2 | 90 | 104 | |
CR761245.2 | - | 1 - 2 | 83 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |