Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203932.1 | NP_989263.1 | 11 - 12 | 1582 | 80 | |
XM_012969824.2 | XP_012825278.1 | 11 - 12 | 1879 | 80 | ||
GenBank mRNA | BC064248.1 | - | 11 - 12 | 1582 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |