Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204002.1 | NP_989333.1 | 2 - 3 | 507 | 76 | |
GenBank mRNA | BC064150.1 | - | 2 - 3 | 507 | 76 | |
CT027966.2 | - | 2 - 3 | 343 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |