Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204007.1 | NP_989338.1 | 3 - 4 | 472 | 80 | |
GenBank mRNA | BC064274.1 | - | 3 - 4 | 472 | 80 | |
CR942724.2 | - | 3 - 4 | 458 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |