Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005621.1 | NP_001005621.1 | 4 - 5 | 586 | 58 | |
GenBank mRNA | BC074589.1 | - | 4 - 5 | 586 | 58 | |
CT025365.2 | - | 4 - 5 | 542 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |